BD Phoenixspec Calibrator Kit DFS Item
For measurement of inoculum density for BBL Crystal System and BD Phoenix System
Supplier:  BD 440911
Catalog No. 23025701
Explore available promotions
Description
Description
  • Measures turbidity of microbial suspensions
  • Portable
  • Battery of A/C
Specifications
Specifications
Probe analysis card, lysis solution, buffer solution, substrate solution, reagent cassettes
Calibrator Kit
Measure turbidity of microbial suspension
Each