BD Saf-T-Intima™ IV Catheter Systems
With Y Adapter
Supplier: BD 383336
Description
- Combines needlestick safety with a closed system design
- Results in a virtually bloodless procedure for vascular access
- Made of clinically provenBD Vialon™ biomaterial
Specifications
Vialon Biomaterial | |
20 G | |
Flexible textured wings, needle shield, extension tubing and Y adapter | |
1 in. | |
Single-use | |
Notched |
Pink | |
Virtually bloodless procedure for vascular access, reduces the risks of infiltration, reduces IV restarts due to reduced complications | |
Yes | |
IV Catheter System | |
25/Pk. | |
Sterile |