BD SafetyGlide™ Needles
Needles feature patented Activation-Assist™ Technology for fast and easy, single-handed needle-tip shielding immediately after injection.
Supplier:  BD 305916
 View more versions of this product
Catalog No. 14-826-106
Explore available promotions
Description
Description
  • This technology provides versatility and minimal change in technique
Specifications
Specifications
Black
General and hypodermic use
Yes
Shielding Hyperdermic Needle
Sterile
25 G
Shielding mechanism
1 in.
Regular Bevel