BD SafetyGlide™ Syringes with Needles
BD SafetyGlide™ Needles are designed with Activation Assist™ technology which allows for fast and easy, single-handed needle-tip shielding
Supplier:  BD 305905
 View more versions of this product
Catalog No. 14-826-102
Explore available promotions
Description
Description
This technology provides versatility and minimal change in technique.
  • Needle-tip shielding Activation-Assist Technology incorporated on the needle, rather than the syringe, offers a greater degree of protection from needlestick injuries
  • BD Luer-Lok™ connection securely attaches SafetyGlide needles to any standard syringe
Order Info
Replacement needles available (See Cat. Nos. 14826106, -07 and 1482910, -11, -13).
Specifications
Specifications
23 G
1 in.
3 mL
Regular Bevel
Detachable subcutaneous injection needle
SafetyGlide™ Syringe
38.1 mm
Luer Lock