BD Sterile Pack Swabs for Environmental Monitoring
BD Sterile Filled Swabs are double-wrapped, gamma-irradiated, ready-to-use sterile swabs for surface sampling, combined with a rinse solution-filled tube.
Supplier:  BD 220518
Catalog No. B220518
Explore available promotions
Description
Description
  • For environmental monitoring
  • Ready-to-use
  • Gamma-irradiated double-pouch design
Specifications
Specifications
Sterile
Tube
Filled Swab Tube
Pre-filled rinse isotonic solution in tube
For environmental monitoring
Product Suggestions
Product Suggestions