BD Tuberculin Syringes
Sterile, disposable syringe with Luer slip tip
$49.66 - $50.55
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Needle Gauge | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Needle Gauge | Price | Quantity | |||
14-826-88
|
BD
309626 |
0.62 in. | 1.6 cm | 25 G |
N/A
|
|
|||
14-829-10D
|
BD
309624 |
1 in. | 2.54 cm | 21 G |
N/A
|
|
|||
14-826-87
|
BD
309623 |
0.5 in. | 1.3 cm | 27 G |
N/A
|
|
|||
Description
- Color-coded needle package and hub In Easy Peel™ package; autoclavable out of package Available with or without detachable PrecisionGlide™ needle
- Syringe 14-823-2F accepts PrecisionGlide, Yale™, and Perfektum™ disposable and reusable needles
Specifications
1 mL |