BD Vacutainer™ CPT™ Mononuclear Cell Preparation Tube
For the one-step isolation of lymphocytes, monocytes, and plasma
Supplier:  BD 362760
Catalog No. 02-688-81
Explore available promotions
Description
Description
  • During centrifugation, a gel barrier in the tube isolates the mononuclear cells and plasma from the erythrocytes and granulocytes
  • May also be used to obtain highly concentrated mononuclear cells and/or plasma for immunotyping, culture, and cell function studies, and for tests using gene-amplification techniques
  • Mononuclear cells may be resuspended in plasma to retain viability for storage and transportation
  • Volume: 4 mL
Specifications
Specifications
Glass
For the separation of mononuclear cells from whole blood
Cell Preparation Tube
4 mL