BD Vacutainer™ Glass Blood Collection Tubes with Sodium Heparin: Hemogard™ Closure
Evacuated glass blood collection tubes for plasma determinations in chemistry
Supplier:  BD 367671
Catalog No. 02-688-8
Explore available promotions
Description
Description
  • Evacuated blood collection tubes with heparin are used for plasma determination in chemistry
  • Tubes are sterile
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Safety shield designed for safer tube handling
  • Rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
  • Shield is designed to protect lab personnel from direct contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube
Specifications
Specifications
13 x 75 mm; Capacity: 2.0 mL
Hemogard
Green
Glass
2 mL
13 x 75 mm