BD Vacutainer™ Glass Blood Collection Tubes with SPS for Microbiology Testing
Evacuated blood culture specimen collection tubes for whole blood microbiology testing
Supplier:  BD 364960
Catalog No. 02-684-19
Explore available promotions
Description
Description
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Yellow color-coded stoppers to enhance speed and accuracy in the laboratory
Specifications
Specifications
Glass
Yellow
Paper
8.3 mL
Microbiology Testing
8.3mL