BD Vacutainer™ Glass Mononuclear Cell Preparation (CPT) Tubes
Evacuated tube intended for the collection of whole blood and the separation of mononuclear cells
Supplier:  BD 362761
 View more versions of this product
Catalog No. 02-685-125
Explore available promotions
Description
Description
BD Vacutainer™ Mononuclear Cell Preparation Tube (CPT™)
  • The one-step, closed-system tube for blood collection, mononuclear cell separation and transportation
  • Contributes to increased lab productivity by offering convenience, safety and reproducibility
  • CPT Cell Preparation tubes are an evacuated blood collection tube system containing either 0.1 M sodium citrate anticoagulant or sodium heparin anticoagulant
  • Tubes also contain blood separation media composed of a thixotropic polyester gel and a FICOLL™ Hypaque™ solution
Specifications
Specifications
Sodium Citrate
Traditional Cap
Light Blue/Black
Mylar (Transparent)
Glass
8 mL
16 x 125 mm