BD Vacutainer™ Safety-Lok™ Blood Collection Sets
Slim, easy-to-handle set allows needle to be easily and completely retracted into the translucent plastic shield and securely locked in place after use
Supplier:  BD 367297
 View more versions of this product
Catalog No. 02-664-7
Explore available promotions
Description
Description
  • Large wing size permits improved positioning during use
  • Multisample Luer adapter (MSLA) allows more than one sample to be taken from a single venipuncture
Compliance
Protective shield is designed to meet current OSHA safety standards for engineering controls.
Specifications
Specifications
Blood Collection Set
23 G
0.75 in.
PVC
OSHA
12 in. without Luer Adapter