BD Vacutainer™ Safety-Lok™ Blood Collection Sets
Slim, easy-to-handle set allows needle to be easily and completely retracted into the translucent plastic shield and securely locked in place after use
Supplier: BD 367298
Description
- Large wing size permits improved positioning during use
- Multisample Luer adapter (MSLA) allows more than one sample to be taken from a single venipuncture
Compliance
Protective shield is designed to meet current OSHA safety standards for engineering controls.
Specifications
Blood Collection Set | |
25 G | |
0.75 in. |
PVC | |
OSHA | |
12 in. without Luer Adapter |