missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Vacutainer™ Urine Collection Kits: Urinalysis
Reduce the need to retest, relabel, and/or recollect specimens
Supplier: BD 364989
Description
- Closed systems that benefit healthcare workers by reducing their need to come into contact with potentially hazardous specimens
- Designed to provide safety and satisfaction, as well as consistent, reliable results to ensure proper patient outcomes
Urinalysis Products
- Include BD™ proprietary mercury-free Urinalysis Preservative Tube, which provides compliance with both the Memorandum of Understanding for the U.S. EPA and the AHA
- Help to maintain urine sample integrity for up to 72 hours at room temperature
- Minimum and maximum fill lines (7-8mL) ensure proper additive-to-urine ratio
Specifications
Urine Collection Kit | |
Sterile | |
Evacuated conical bottom tube and cup | |
Room temperature | |
8 mL |
Plastic | |
For UA chemistry Urine testing | |
Yes | |
0.27 oz. |