missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Vacutainer™ Urine Collection Kits: Urinalysis
Reduce the need to retest, relabel, and/or recollect specimens
Supplier: BD 364946
Description
- Closed systems that benefit healthcare workers by reducing their need to come into contact with potentially hazardous specimens
- Designed to provide safety and satisfaction, as well as consistent, reliable results to ensure proper patient outcomes
Urinalysis Products
- Include BD™ proprietary mercury-free Urinalysis Preservative Tube, which provides compliance with both the Memorandum of Understanding for the U.S. EPA and the AHA
- Help to maintain urine sample integrity for up to 72 hours at room temperature
- Minimum and maximum fill lines (7-8mL) ensure proper additive-to-urine ratio
Specifications
| Urine Collection Kit | |
| Sterile | |
| For UA chemistry Urine testing | |
| Yes | |
| 0.27 oz. |
| Plastic | |
| Conical | |
| Preservative, Transfer Device, Tube, Cup | |
| Room temperature | |
| 8 mL |