BD Vacutainer™ Venous Blood Collection Tubes: Vacutainer Plus™ Glass Serum Tubes, Silicone-Coated, with Conventional Stopper
Use for serum determinations in chemistry, serology and immunohematology
Supplier: BD 366430
Description
- Sterile vacuum blood collection tubes for serum determination
- Spray-coated silica interior
- With conventional tube stopper
Evacuated Tubes Simplify Blood Collection
- Blood is drawn directly from the vein into the evacuated sterile collection tubes
- Only one venipuncture is needed to draw multiple samples
- Tubes can be changed while needle and plastic holder remain in place on patient's arm
- Closures are color coded
Specifications
Glass | |
10 mL | |
16 x 100 mm |
Traditional Cap | |
Red |