missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Vacutainer™ Venous Blood Collection Tubes: Vacutainer Plus™ Plastic Serum Tubes, Silicone-Coated, with Conventional Stopper
Use for serum determinations in chemistry, serology and immunohematology.
$41.61 - $47.50
Specifications
Additive | Clot Activator, Silicone Coated |
---|---|
Material | Plastic |
Closure Type | Traditional Cap |
Closure Color | Red |
Catalog Number | Mfr. No. | Diameter (Metric) Outer | Capacity (Metric) | Description | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Outer | Capacity (Metric) | Description | Price | Quantity | |||
02-657-27
|
BD
366668 |
13 x 75 mm | 3 mL | Plastic Serum Tube |
N/A
|
|
|||
02-683-94B
|
BD
367820 |
16 × 100 mm | 10 mL | BD Vacutainer™ Plus plastic serum tube |
N/A
|
|
|||
Description
- Sterile vacuum blood collection tubes for serum determination
- Spray-coated silica interior
- Tubes marked as Best Practice Formulary meet demanding requirements of complex diagnostic instruments and current clinical guidelines
- With conventional tube stopper
Evacuated Tubes Simplify Blood Collection
- Blood is drawn directly from the vein into the evacuated sterile collection tubes
- Only one venipuncture is needed to draw multiple samples
- Tubes can be changed while needle and plastic holder remain in place on patient's arm
- Closures are color coded
- Made of polyethylene terephthalate (PET)
- Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; (2) eliminating breakage when tubes are centrifuged at higher speeds; and (3) eliminating breakage or chipping of tube rims when stopper is removed and reinserted
- In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation
Specifications
Clot Activator, Silicone Coated | |
Plastic | |
Traditional Cap | |
Red |