BD Vacutainer™ Venous Blood Collection Tubes: Vacutainer Plus™ Plastic Serum Tubes, Silicone-Coated, with Hemogard™ Closure
Use for serum determinations in chemistry, serology and immunohematology
Supplier:  BD 367814
 View more versions of this product
Catalog No. 02-687-94
Explore available promotions
Description
Description
  • Sterile vacuum blood collection tubes for serum determination
  • Spray-coated silica interior
  • With Hemogard™ tube closure, which prevents human contact with blood on the stopper or tube rim
Evacuated Tubes Simplify Blood Collection
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded
Plastic Evacuated Tubes
  • Made of polyethylene terephthalate (PET)
  • Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; (2) eliminating breakage when tubes are centrifuged at higher speeds; and (3) eliminating breakage or chipping of tube rims when stopper is removed and reinserted
  • In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation
Hemogard™ Stoppers
  • Safety shield designed for safer tube handling
  • Rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
  • Shield is designed to protect lab personnel from direct contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube
Specifications
Specifications
Clot Activator, Silicone Coated
Hemogard
Red
Plastic
5 mL
13 x 100 mm