BD Vacutainer™ Winged Safety Push Button Blood Collection Sets without Luer Adapter
Blood collection set offers in-vein safety activation provides split-second protection against accidental needlestick injuries
Supplier: BD 367323
Description
- Ease of use reduces training requirements and improves activation compliance
- In-vein activation reduces the risk exposure to a contaminated needle
- Easy activation without additional patient discomfort
- Ideal for use in high-risk environments No luer adapter
Specifications
Push Button Blood Collection Set | |
25 G | |
3/4 in. | |
12 in. |