BD Veritor™ Plus System
The BD Veritor™ Plus System enhances point-of-care testing for SARS-CoV-2,Flu A+B, RSV and Group A Strep assays, providing rapid diagnostic testing in a convenient, portable instrument.
Supplier: BD B256066
Description
- Improved workflow with two modes - Analyze Now and Walk-Away mode providing test results and aiding in decision making while the patient is onsite
- Cleared for use with nasal swabs and nasopharyngeal swabs
- Reduce hands on time
- Create a flexible workflow
- Improved traceability with InfoScan module
- Better connectivity with InfoSync module
- Advanced particle technology enhances sensitivity by using a proprietary process to produce highly stable, modified colloidal metal particles
- Adaptive read technology compensates for background and non-specific binding which helps improve specificity to reduce false-positive results
- Number of reads per instrument: 3500
- Rechargeable lithium battery
- Print-enabled
Specifications
BD Veritor™ Point-of-Care Immunoassay Analyzer | |
Influenza A and B, Respiratory Syncytial Virus (RSV), Streptococcus (Group A) | |
Point-of-Care ImmunoAssay Analyzer | |
Lithium battery | |
5 to 10 min. |
Waived | |
Nasal swabs and nasopharyngeal (NP) swabs | |
InfoSync Module | |
Each |