BD Veritor System RSV (Respiratory Syncytial Virus) Test Kit
Provides objective results on an easy to read digital display, unlike current rapid RSV tests, which requires subjective visual test interpretation to determine positive or negative test result
$541.59 - $541.59
Specifications
Detectable Analytes Respiratory Syncytial Virus (RSV)
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
B256038
View Documents
BD
256038
For use in point of care settings
Pack of 30 Each for $541.59
B256042
View Documents
BD
256042
For laboratory use
Pack of 30 Each for $541.59
Description
Description
  • Intended for in vitro diagnostic use to aid in diagnosis of RSV infections in infants and pediatric patients under age of 20 years
  • For use with BD Veritor System Reader
Specifications
Specifications
Respiratory Syncytial Virus (RSV)
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'