HemoCue America Beckman Coulter™ ICON™ 20 hCG Urine/Serum Combo Test
Simple, one-step process for greater reliability and higher sensitivity
Supplier:  HemoCue America 395097A
Catalog No. 23-280-014
Explore available promotions
Includes: 25 test devices, disposable droppers, product instructions
Description
Description
  • CLIA complexity: Waived for urine; Non-waived for serum
  • CPT codes: 81025 (urine); 84703 (serum)
  • Sensitivity: Detects hCG at 10mIU/mL in serum and 20mIU/mL in urine
  • Solid-phase chromatographic immunoassay selectively detects elevated levels of hCG in serum or urine
  • Results in three to five minutes
  • Built-in positive and negative controls
  • Requires just three drops of sample
  • Long shelf life
Specifications
Specifications
81025, 84703
hCG
Combo Test Kit
25 Tests
3 min. for urine, 5 min. for serum
Waived, Non-waived
Serum, Urine
25 Test devices, disposable droppers, product instructions
3 Drops