missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Beclomethasone dipropionate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 460992500

 View more versions of this product

Catalog No. AC460992500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Beclomethasone dipropionate
1% max. (105°C, 3 h)
96% min. (HPLC)
250mg
KUVIULQEHSCUHY-XYWKZLDCSA-N
[2-[(8R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
134129500
97%
5534-09-8
Authentic
C28H37ClO7
beclomethasone dipropionate, beclometasone dipropionate, beconase, beclovent, vancenase, beclazone, becloforte, beclomet, beclorhinol, propaderm
CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)Cl)O)C)C)OC(=O)CC
521.04
521.04

Chemical Identifiers

5534-09-8
521.04
beclomethasone dipropionate, beclometasone dipropionate, beconase, beclovent, vancenase, beclazone, becloforte, beclomet, beclorhinol, propaderm
[2-[(8R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
C28H37ClO7
KUVIULQEHSCUHY-XYWKZLDCSA-N
134129500
CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)Cl)O)C)C)OC(=O)CC
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 226-886-

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only