missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
High-Tech Conversions Bee-Safe™ Cleanroom Bouffant Caps
High-Tech Conversions Bee-Safe™ Cleanroom Bouffant Caps are breathable, and comfortable during periods of long-wear
Supplier: High-Tech Conversions GAHBP28WH
Description
- Perfect for compact storage and easy dispensing
- Soft & double-stitched for more comfort, breathable for long periods of wear
- High quality, economically priced
- For use in controlled environments, food prep/processing, and industrial settings
Specifications
Cleanroom Bouffant | |
Polypropylene | |
Yes | |
Non-sterile | |
24 in. | |
24 in. |
28 in. | |
Single-use | |
24 in. | |
100/Pk. | |
28 in. | |
Controlled Environments, Food Prep/Processing, and Industrial Applications |