missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Behenic Acid 95.0+%, TCI America™

Supplier:  TCI America B12485G

 View more versions of this product

Catalog No. B12485G


Explore available promotions

Description

Description

Specifications

Specifications

Behenic Acid
80°C
306°C
MFCD00002807
behenic acid, 1-docosanoic acid, n-docosanoic acid, hydrofol acid 560, docosoic acid, hydrofol 2022-55, glycon b-70, behensaeure, docosanoicacid, docosansaeure
CCCCCCCCCCCCCCCCCCCCCC(=O)O
340.592
CHEBI:28941
≥95.0% (GC)
112-85-6
White-Yellow
C22H44O2
5 g
UKMSUNONTOPOIO-UHFFFAOYSA-N
docosanoic acid
8215
340.59
Crystalline Lumps

Chemical Identifiers

112-85-6
340.592
UKMSUNONTOPOIO-UHFFFAOYSA-N
8215
docosanoic acid
C22H44O2
MFCD00002807
behenic acid, 1-docosanoic acid, n-docosanoic acid, hydrofol acid 560, docosoic acid, hydrofol 2022-55, glycon b-70, behensaeure, docosanoicacid, docosansaeure
CHEBI:28941
CCCCCCCCCCCCCCCCCCCCCC(=O)O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0608

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions