Bel-Art™ SP Scienceware™ Acid/Solvent Bottle Carriers
Polyethylene carriers with epoxy coated, heavy-gauge steel wire bails have wide ribs in both the sides and on the bottom to support and cushion bottles
Supplier:  Bel-Art™ F169580000
 View more versions of this product
Catalog No. 22-260395
Explore available promotions
Description
Description
  • Offer shatter resistant protection for glass bottles
  • Cone style lid has a 5/8 in. (16mm) opening in top for insertion of tubing; simplifies the transfer of liquids without pouring
  • Truncated style lid permits pouring without the removal of the lid
  • Designed to hold standard US reagent bottles
Specifications
Specifications
Bottle Carrier
Bottles (4 L)
37 cm
Not Autoclavable
Polyethylene
14.37 in.