missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Antibody Saver Trays
Minimize the use of expensive antibodies during incubations by using an optimally sized tray
Supplier: Bel-Art™ F451000000
Description
- For popular midi and mini gels
- Stackable, reclosable and reusable
- All trays include attached lid to protect against spillage
- Single lane trays are supplied as a group of five joined lanes; can be easily separated to single lane prior to use
- Polypropylene; autoclavable
Specifications
0.62 x 4.5 in. (1.6 x 11.5 cm) | |
Polypropylene | |
11.5 cm | |
0.625 in. | |
4.5 in. |
1.6 cm | |
Antibody Saver Tray | |
Attached lid to protect against spillage | |
Rectangular | |
Autoclavable |