missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Antibody Saver Trays
Minimize the use of expensive antibodies during incubations by using an optimally sized tray
Supplier: Bel-Art™ F451000001
Description
- For popular midi and mini gels
- Stackable, reclosable and reusable
- All trays include attached lid to protect against spillage
- Single lane trays are supplied as a group of five joined lanes; can be easily separated to single lane prior to use
- Polypropylene; autoclavable
Specifications
3.5 x 3.5 in. (9 x 9 cm) | |
Polypropylene | |
9 cm | |
3.5 in. | |
3.5 in. |
9 cm | |
Antibody Saver Tray | |
Attached lid to protect against spillage | |
Square | |
Autoclavable |