missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Benchtop Biohazard Disposal Bags
For disposal of tips and tubes.
Supplier: Bel-Art™ F131660000EMD
Description
- Bright red
- Features one long flap for easy opening
- Printed with biohazard symbol and sterilization indicator patch
- Made from high molecular weight high-density polyethylene
- Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
- Size allows bag to be inverted over hand to safely pick up waste
- Steam autoclavable at 121°C (250°F)
Specifications
Benchtop Biohazard Disposal Bag | |
1627.7 mL | |
Red with Black Print | |
For disposal of tips and tubes. | |
HDPE | |
0.72 mil | |
22 cm | |
28 cm |
Autoclavable | |
Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests | |
Single-use | |
Printed biohazard symbol and sterilization indicator patch, one long flap | |
100 | |
8.5 in. | |
11 in. |