missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Benchtop Biohazard Waste Disposal Bags, Bright red; W x H: 8.5 x 11 in. (22 x 28cm); 100 per pack

Bright red, autoclavable bags feature one long flap for easy opening and disposal of tips and tubes

Supplier:  Bel-Art™ 131660000

Catalog No. 03-410-591


Explore available promotions

Description

Description

Bright red, autoclavable bags feature one long flap for easy opening and disposal of tips and tubes

  • Size allows the bag to be inverted over user's hand to safely pick up waste
  • Printed with the biohazard symbol and sterilization indicator patch
  • Constructed from High Molecular Weight, High-Density (HMHD) polyethylene
  • Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
  • Steam autoclavable at 121°C (250°F)
  • For space-saving waste disposal, use with Poxygrid™ Benchtop Biohazard Bag Holder (Mfr. No. H13193-1000)

 


Specifications

Specifications

Printed Biohazard Symbols
Single-use
22 cm
Easy Opening and Disposal of Tips and Tubes
0.07 mm
HDPE
Red
28 cm
Autoclavable
8.5 in.
11 in.
Benchtop Biohazard Waste Bag
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions