Bel-Art™ SP Scienceware™ Bent Handle Scoop
Features an ergonomic, curved handle design to elevate the hand from the sample, reducing risk of contamination
Supplier:  Bel-Art™ H369022006
 View more versions of this product
Catalog No. 14-241-202
Explore available promotions
Description
Description
  • Curved handle helps prevent incidental contact with sample source material and exposure to hazardous substances
  • Flat bottom keeps scoop upright when placed on a flat surface; can be used as a weigh boat
  • Made from high-impact, lightweight, FDA grade polystyrene
  • Sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Sterilization
  • Individually packaged and ready for one time use and disposal
Specifications
Specifications
Scoop
Flat
2 oz.
Single-use
FDA
7.62 x 1.62 x 1.12cm (19.4 x 4.1 x 2.9 cm)
1.12 in.
2.9 cm
3.06 in.
19.4 cm
11.6 cm
1.62 in.
4.1 cm
Polystyrene
Bent and curved
60 mL
Sterile
Design to elevate the hand from the sample, reducing risk of contamination, prevent incidental contact with sample source material and exposure to hazardous substances
1.12 in.
2.9 cm
7.62 in.
4.56 in.
7.62 cm
1.62 in.
4.1 cm