missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ Clavies™ Heat-Resistant Biohazard Autoclave Gloves
Soft, pliant, cotton terrycloth gloves are ideal for handling hot objects out of the autoclave or oven and protect from temperatures up to 450°F (232°C).
Supplier: Bel-Art™ H132010000
Description
Soft, pliant, cotton terrycloth gloves are ideal for handling hot objects out of the autoclave or oven and protect from temperatures up to 450°F (232°C).
- Ideal for removing hot objects from an autoclave or oven
- Heat-resistant up to 232°C
- Replaces hazardous asbestos gloves
- Extended gauntlet cuff available for additional forearm protection
- Gloves are not waterproof and should not be used to handle dry ice
- Machine wash, air dry, or use the Lab-Aire™ II Glove Holder (F18931-0000) to dry on any Lab-Aire™ dryer
Specifications
CE | |
Laboratory, Industrial | |
Orange | |
Terrycloth | |
Universal | |
450°F | |
Handling Hot Objects | |
H13201 | |
5 in. | |
13 cm |
Yes | |
Uncoated | |
Yes | |
Autoclave Gloves | |
Full-hand Mitt | |
232°C | |
Gauntlet | |
13 in. | |
33 cm |