missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ Clavies™ Biohazard Bag Holders
Keep bags open and ready to use
Supplier: Bel-Art™ F131920001
Description
- Economical, lightweight stainless-steel wire frame
- Highly visible orange polypropylene bottom tray for easy identification as a biohazard containment system
- Tray serves as containment well in case of bag leakage
- Autoclavable at 121°C (250°F) and dishwasher safe
- Assembly is easy and permanent
- Medium and large models can be used with autoclavable polypropylene cover(Mfr. No. F13192-0102) to improve sanitation
Specifications
Biohazard Bag Holder | |
Silver, Orange | |
Designed to securely support the bag and its contents | |
Stainless steel wire frame and orange polypropylene bottom, plastic wire anchors | |
Sterile | |
Small | |
61 cm | |
47 cm |
Autoclavable | |
Reusable | |
01-826A, 01-826-5, 01-827B, 01-814-1, 01-815A small 30 x 61cm Clavies™ biohazard bag | |
Stainless Steel | |
Benchtop | |
24 in. | |
18.5 in. |