missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Conical Tube Holder

Allows user to clearly view the entire contents of tubes. Bel Art™ SP Scienceware™ Conical Tube Holder sets up in just seconds to hold either a 15ml or a 50ml conical tube.

Supplier:  Bel-Art™ F187960000

Catalog No. 11-999-001


Explore available promotions

Description

Description

Allows user to clearly view the entire contents of tubes. Bel Art™ SP Scienceware™ Conical Tube Holder sets up in just seconds to hold either a 15ml or a 50ml conical tube.

  • Versatile holder supports either a 15 or 50mL conical tube while allowing a view of the entire tube contents
  • Move tube and rack with one hand by pinching the uprights
  • Insert or remove tube from the holder with one hand
  • Steam autoclavable at 121°C (250°F)
  • Chemical-resistant polypropylene

Specifications

Specifications

Test Tube Single Rack
Transparent
Polypropylene
15 and 50mL Conical Tubes
Autoclavable
Tubes (15 or 50 mL)
1
Videos
SDS
Documents

Documents

Promotions

Promotions