Bel-Art™ Earth Friendly 1001 Sticks
Lessen environmental impact with polylactic acid (PLA) resin sampling sticks
$234.29 - $234.29
Specifications
Autoclavable Not Autoclavable
Shape Blunt End
Sterility Non-sterile
Certifications/Compliance FDA
Product Type Earth-Friendly™ Sampling Stick
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
14-512-205
View Documents
Bel-Art™
F369310000
100 Pk.
Pack of 100 Each for $234.29
Description
Description
  • The production of 100% plant-based PLA contributes fewer greenhouse gases and uses less nonrenewable energy from source to resin than its petroleum-based counterparts.
  • Plants used are an annually renewable resource.
  • Suitable for mixing samples, handling capsules, scraping containers or positioning solid samples for examination.
  • Green alternative to petroleum-based sampling tools helps meet sustainability goals.
  • Comparable in strength to polystyrene, polypropylene and other petroleum-based plastic resin sampling tools.
  • Disposal options include industrial composting for biodegradability, incineration, recycling and landfill.
  • Nonsterile FDA-grade: safe for food, drug and cosmetic contact.
  • Use a separate stick for each material sampled to avoid cross-contamination.
Specifications
Specifications
Not Autoclavable
Non-sterile
Earth-Friendly™ Sampling Stick
5.75 in.
1.9 cm
PLA Resin
Blunt End
FDA
Suitable for mixing samples, handling capsules, scraping containers or positioning solid samples for examination
0.75 in.
5.75 x 0.75 in. (14.6 x 1.9 cm)
14.6 cm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'