Bel-Art™ SP Scienceware™ Earth Friendly Long-Handle Spoons
Lessen environmental impact with PLA (polylactic acid) resin sampling spoons
Supplier:  Bel-Art™ F369401000
 View more versions of this product
Catalog No. 14-512-207
Explore available promotions
Description
Description
The production of 100% plant-based PLA contributes fewer greenhouse gases and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource. Disposal options include industrial composting for biodegradability, incineration, recycling and landfill.
  • FDA-grade spoons are safe for food, drug and cosmetic contact
  • Comparable in strength and durability to polystyrene, polypropylene and other plastic resin sampling tools
  • Long handle, 7 in. (17.8cm), helps prevent personal contact with sampled material when reaching into bottles, jars and other confined areas
  • Rim of each bowl has a flat edge to assist in collecting and measuring small amounts with accuracy
  • Use a separate spoon for each material sampled to avoid cross contamination
  • Nonsterile
Specifications
Specifications
Earth Friendly Sampling Spoon
Flat edge rim
1.23 mL
FDA
7 in.
PLA Resin
Long
Non-sterile
Ideal for acquiring samples from bottles, jars and other confined areas and helping to avoid personal contact with sample materials, used as a green alternative to petroleum-based sampling tools
17.8 cm