Bel-Art™ SP Scienceware™ Economy Dippers
For convenient sampling from tanks, vats, streams, ponds, lakes
Supplier:  Bel-Art™ F367860016
 View more versions of this product
Catalog No. 03-410-811
Explore available promotions
Description
Description
  • Lightweight and portable
  • Thread on 500mL (16 oz.) polypropylene cup can be exchanged to reduce cross contamination
  • Two pouring spouts on cup allow left or right pouring
  • High-density polyethylene plastic handles are strong honeycomb tubes without inserts and are ribbed for sure grip
  • Handle end has a hook for hanging storage
Specifications
Specifications
Dipper Cup
Autoclavable
500 mL
HDPE
16 oz.
For sampling from tanks, ponds, lakes