H-B Instrument™ Five-Hole Stainless-Steel Clamping Thermometer Holder
Securely hold thermometers. Bel-Art™ SP Scienceware™ Five-Hole Stainless-Steel Clamping Thermometer Holder is spring loaded with rubber bumpers on a clip.
Catalog No. 13-204-000
Explore available promotions
Description
Description
With five hole sizes ranging from 3.5mm to 14.5mm, this holder accommodates various thermometers including liquid in glass, electronic with probe, and bimetal with probe.
  • Stainless steel, spring loaded clip attaches securely to laboratory containers, shelves up to 3/8 in. thick, and more
  • Rubber bumpers on clip prevent scratching
  • Durable PE foam provides secure grip to keep thermometers in place
  • Dimensions: 8.3cmL x 5.4cmW x 6.4cmH (3.25 x 2.13 x 2.5 in.)
Specifications
Specifications
Stainless-steel spring-loaded clip, rubber bumpers on clip ends, polyethylene foam thermometer holder
Five Hole
3.25 in.
83 mm
Thermometer Holder
Thermometers