Bel-Art™ SP Scienceware™ Hand-Held Colony Counter
Bel-Art™ SP Scienceware™ Hand-Held Colony Counter is simple-to-operate colony counter marks, counts and gives an audible and visual alert each time count is registered.
Supplier:  Bel-Art™ F378620002
Catalog No. 07-910-018
Explore available promotions
Includes: Long-life red and black felt-tip markers, 3V lithium battery, protective storage case.
Description
Description
  • Fast, accurate and economical sample counting and sample count verification
  • Stores up to 35 sample counts
  • Verifies sample counts with up/down counting
  • Displays sum total of all colonies counted
  • Backlit display for easy viewing in low light conditions
  • Audible alert can be turned off if desired
  • Counting function can be locked to prevent error during breaks between counting
  • Included felt-tip markers work on plastic or glass and are oil- and water-resistant; ink is removable with ethyl alcohol
Order Info
Colony Counter System (07-910-022) also available. Replacement markers (07-910-019 and 07-910-020) and battery (07-910-021) sold separately.
Specifications
Specifications
Colony Counter
Stores up to 35 samples
CE
Long life red and black felt tip markers, 3V Lithium battery, protective storage case
Backlit LCD
6 in.
Count colonies of Bacteria or other microorganisms grown on an agar plate