Bel-Art™ Lab-Aire™ II Glassware Drying Racks
Glassware and plasticware drying racks for walls or benchtops
Supplier:  Bel-Art™ F189330022
 View more versions of this product
Catalog No. 03-422-200
Explore available promotions
Description
Description
Glassware and plasticware drying racks for walls or benchtops
  • Dry and store delicate labware including beakers, flasks, bottles, test tubes and cylinders on these space-saving drying racks
  • Single-sided benchtop models fit flat against a wall
  • Double-sided benchtop models provide double the capacity (up to 114 pegs) with a slightly larger footprint
  • Three sizes and lengths of V-channel pegs can be repositioned to hold a variety of labware
  • Rugged ABS plastic body and pegs won't corrode and wipe clean
  • Wall-mount models have mounting points that fit standard pegboards
  • Pegs available separately to reconfigure rack for specific drying requirements
Specifications
Specifications
Benchtop Rack (2-tier)
ABS
37.5 cm
39 cm
10 in.
Lab Glassware, Lab Plasticware
76
14.75 in.
15.4 in.
25.4 cm