missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Micro Funnels
Designed for micro work, filling burets, manometers and small bottles
Supplier: Bel-Art™ F146850028
Description
- Polypropylene funnels are steam autoclavable at 121°C (250°F)
Specifications
5.2 mL | |
Polypropylene | |
For micro work, filling burettes, manometers and small bottles |
0.6cm | |
Autoclavable |