Bel-Art™ SP Scienceware™ Micro Spoon and Spatula Sampling Set
Includes six different, small-size, double-ended tools and tweezers for accurate handling of small quantities. Bel Art™ SP Scienceware™ Micro Spoon and Spatula Sampling Set are polished stainless steel for long life and easy cleaning and can be steam autoclaved at 121ºC (250ºF)
Supplier:  Bel-Art™ F367050000
Catalog No. 11-865-130
Explore available promotions
Description
Description
Includes six different, small-size, double-ended tools and tweezers for accurate handling of small quantities. Bel Art™ SP Scienceware™ Micro Spoon and Spatula Sampling Set are polished stainless steel for long life and easy cleaning and can be steam autoclaved at 121ºC (250ºF)
  • Each tool has two different end shapes, doubling the selection of implements to better match user needs
  • Tool ends include 2 small spoons for transferring and collecting both dry and liquid material
  • Eight flat spatulas of various widths and lengths excellent for mixing, scraping and spreading; Six of these have a square end and two are rounded
  • Two V-shaped spatulas of different sizes can be used for lifting or spill-free carrying of material
  • The blunt-tip tweezers have serrated inside surfaces for secure sample handling and grooved handles for better gripping
  • All tools are polished stainless steel for long life and easy cleaning and are steam autoclavable at 121°C (250°F)
  • A carrying case protects and organizes all tools when not in use
Specifications
Specifications
Micro Spoon and Spatula Sampling Set
Autoclavable
Reusable
Stainless Steel
6 small size double-ended tools, set of tweezers, 2 small spoons, 6 square end flat spatulas, 2 round end flat spatulas, 2 v-shaped spatulas, blunt tip tweezers, carrying case
Transferring and collecting both dry and liquid material