Learn More
Bel-Art™ Pipet Vacuum Aspirator Collection System
Performs vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes.
Supplier:  Bel-Art™ F199170150
 View more versions of this product
Catalog No. 13-717-260
Only null left
Explore available promotions
Description
Description
  • Built-in pump quickly aspirates fluids from pipet tip, Pasteur pipet or serological pipet inserted into the collection tube
  • Maintains low noise level (76 db at 1 meter) and minimal vibration
  • Excellent for fluid collection from tissue and/or cell culture, Petri dishes, plates, flasks and tubes
  • Chemical and corrosion resistant polypropylene collection bottle holds 3800 mL (1 gallon) fluid wastes and has a safety release vent to prevent vacuum implosion
  • A replaceable hydrophobic filter seals itself if it becomes wet protecting the pump motor from fluid contamination
  • Lightweight and compact design will not crowd workspace in a hood or at the bench
Specifications
Specifications
Pipettes
Pipet Vacuum Aspirator Collection System
Media Bottles
Plastic
3800 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories