missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ Bench-Top Biohazard Bag Holder
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
Supplier: Bel-Art™ F131930102
Includes: Plastic Cover Only
Description
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
- Rectangular steel wire frame to hold one polyethylene bag of dimensions 8.5 x 11 in. (22 x 28cm)
- Available with epoxy coat in orange color
- Comes with snug fitting polypropylene plastic cover to hold bag securely in place
- Reusable and allows cleaning by hand or in automated dish washers
- Autoclavable
- Also used with Clavies Autoclavable Bags H13182-0812
Order Info
For complete kit with bag holder, cover and biohazard bags, see Cat. No. 11-394-310.
Specifications
Bag Holder Cover | |
Orange | |
Store, transport, or ship biohazardous wastes | |
Snug-Fitting Plastic Cover Only | |
5 in. |
Autoclavable | |
Single-use | |
Biohazard bag holder | |
Polypropylene | |
12.7 cm |