Bel-Art™ SP Scienceware™ Bench-Top Biohazard Bag Holder
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
Supplier:  Bel-Art™ F131930500
 View more versions of this product
Catalog No. 11-394-310
Explore available promotions
Includes: Cover, Bag Holder, and 100 Bench Top Biohazard Bags, made of high molecular weight, high-density (HMHD) Polyethylene and printed with Biohazard symbol and sterilization indicator patch that darkens when exposed to steam sterilization.
Description
Description
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
  • Rectangular steel wire frame to hold one polyethylene bag of dimensions 8.5 x 11 in. (22 x 28cm)
  • Available with epoxy coat in orange color
  • Comes with snug fitting polypropylene plastic cover to hold bag securely in place
  • Reusable and allows cleaning by hand or in automated dish washers
  • Autoclavable
  • Also used with Clavies Autoclavable Bags H13182-0812
Compliance
When used with lid, provides compliance with 29 CFR 1910.1030(d)(2)(xiii) (A), which requires a cover on any container used to store, transport or ship biohazardous wastes.
Specifications
Specifications
Biohazard Bag Holder Kit
Pink
Perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use
Complete Kit: Bag Holder, Cover and Bags (100pk)
Autoclavable
Single-use
01-828A, 01-828-031, 01-830A, 01-826-1, 01-827A, 01-829A, 01-815-1 22 x 28cm biohazard bag, Clavies™ autoclavable bags (01-815-6)
Videos
SDS
Documents
Documents
Accessories