missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ Quick Disconnects

Polyethylene connectors are ideal for vacuum applications, water lines, manometers, and more.

Supplier:  Bel-Art™ F197300002

Catalog No. 22-235663



Explore available promotions

Description

Description

  • Connects same size tubing

Specifications

Specifications

Not Autoclavable
2.6 in.
6.4 to 9.5mm (0.25 to 0.375 in.)
Chrome
Polyethylene
67 mm
Male Fitting: 6.4 to 9.5mm (0.25 to 0.375 in.)
Female Fitting: 6.4 to 9.5mm (0.25 to 0.375 in.)
Quick DisConnect
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions