missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Red Wash Bottles
The bright red color helps identify hazardous contents at a glance
$102.37 - $118.67
Specifications
Color | Red |
---|---|
Material | LDPE |
Autoclavable | Not Autoclavable |
Closure Material | Polypropylene |
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Mouth | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Mouth | Price | Quantity | |||
01-189-0107
|
Bel-Art™
F116500008 |
250 mL | 28 mm | Narrow |
Pack of 6 Each for $102.37
Case of 6 Pack for $614.22
|
|
|||
01-189-0108
|
Bel-Art™
F116500016 |
500 mL | 28 mm | Narrow |
Pack of 6 Each for $111.73
Case of 4 Pack for $446.92
|
|
|||
01-189-0109
|
Bel-Art™
F116510016 |
500 mL | 53 mm | Wide |
Pack of 6 Each for $118.67
Case of 4 Pack for $474.68
|
|
|||
Description
- Low-density polyethylene with polypropylene closure
Specifications
Red | |
LDPE | |
Not Autoclavable | |
Polypropylene |