Bel-Art™ Replacement Shelf and Rails for Dry-Keeper™ Desiccator
Replace or add additional shelves for DryKeeper™ vertical desiccator cabinets.
$97.00 - $305.69
Specifications
For Use With (Application) | Vertical DryKeeper desiccators |
---|---|
For Use With (Equipment) | Vertical DryKeeper desiccators |
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
14-809-302
|
Bel-Art™
H942056602 |
Replacement Shelf |
Each for $305.69
|
|
|||||
14-809-303
|
Bel-Art™
H942053605 |
Replacement Shelf Rail |
Each for $97.00
|
|
|||||
Description
- The Dry-Keeper desiccator cabinet has sleek black ABS accents, vertical profile, and movable shelves for adjusting dry storage space configuration for moisture-sensitive materials
- Adjustable, removable, and perforated shelves facilitate full circulation of air
- Shelf rails can be repositioned on any one of 15 steps set at 28 mm intervals
Specifications
Vertical DryKeeper desiccators | |
Vertical DryKeeper desiccators |