Bel-Art™ SP Scienceware™ Safety Acid/Caustic Sampler
Well-suited for sodium hydroxide, potassium hydroxide, hydrochloric acid, acetic acid, formaldehyde, sulfuric acid, etc.
Supplier: Bel-Art™ H378010000
Description
- Excellent for sampling from tank cars, tank trucks and barges
- A 500cc (16 oz.) polyethylene bottle is screwed onto a 10cm (4 in.) long, 38mm (1.5 in.) diameter bar
- Bar has a slot for fluid entry
- The upper end of the bar is threaded to accept the handle
Specifications
Acid or Caustic Sampler | |
T-shape | |
500 mL | |
3 ft. |
Polyethylene | |
16 oz. | |
Used to sample tanks rail cars and other large vessels | |
0.9 m |