Bel-Art™ SP Scienceware™ Secador™ Auto-Desiccator Module Desiccant Replacement Beads
Color-indicating silica gel beads for use in refilling the Secador auto-desiccator module
Supplier:  Bel-Art™ 420740020
Catalog No. 13-717-26
Explore available promotions
Description
Description
Automatically regenerates for thousands of cycles.
  • Absorbs moisture from the air
  • Inert, non-toxic, non-corrosive
  • Won't disintegrate into powder even after repeated regeneration
  • Color-changing moisture indicator turns from blue to light pink indicating saturation
  • Pink/dark brown hues indicate replacement due