Bel-Art™ SP Scienceware™ Slide Boxes
Will not warp, splinter or crack, and are unaffected by moisture. Bel-Art™ SP Scienceware™ Slide Boxes are compact, durable slide boxes to provide the maximum protection for 25 x 75mm (1 x 3 in.) microscope slides.
$82.47 - $192.97
Specifications
For Use With (Equipment) Microscope Slide
Includes Numbered index on base
View More Specs
Products 2
Catalog Number Mfr. No. Slide Capacity Price Quantity  
Catalog Number Mfr. No. Slide Capacity Price Quantity  
22-261929
View Documents
Bel-Art™
H445770000
100
Each for $192.97
14-512-37
View Documents
Bel-Art™
H445760000
25
Pack of 2 Each for $82.47
Description
Description
  • Heavy-walled boxes are rugged and will not bend, split or chip
  • Boxes are unaffected by humidity and moisture and are thoroughly insect-proof
  • Easily identify slides with numbered index included on base and cover
Specifications
Specifications
Microscope Slide
Numbered index on base
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'